CO1H324A6ADA CombiPhos Catalysts Inc

2-Hydroxythiazole-4-boronic acid pinacol ester

  • Cas Number : 2223051-02-1
  • Molecular Weight : 227.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart