CO1H324A6ADF CombiPhos Catalysts Inc

4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(thiophen-2-yl)thiazole

  • Cas Number : 1402166-57-7
  • Molecular Weight : 293.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart