CO1H324A6B12 CombiPhos Catalysts Inc

2-(4-fluoro-2-methylphenyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiazole

  • Cas Number : 1402227-76-2
  • Molecular Weight : 319.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart