CO1H324A6B1E CombiPhos Catalysts Inc

3-Cyanopyridine-2-boronic acid pinacol ester

  • Cas Number : 1309981-31-4
  • Molecular Weight : 230.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart