CO1H324A6B2D CombiPhos Catalysts Inc

2-(4-Tolyl)pyridine-5-boronic acid pinacol ester

  • Cas Number : 2223028-74-6
  • Molecular Weight : 295.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart