CO1H324A6B31 CombiPhos Catalysts Inc

2-(2-Methoxyphenyl)pyridine-5-boronic acid pinacol ester

  • Cas Number : 2223038-76-2
  • Molecular Weight : 311.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart