CO1H324A6B45 CombiPhos Catalysts Inc

2-(3-Methylpiperidin-1-yl)pyridine-5-boronic acid pinacol ester

  • Cas Number : 2223009-69-4
  • Molecular Weight : 302.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart