CO1H324A6B49 CombiPhos Catalysts Inc

5-(3-methyl-1H-pyrazol-1-yl)-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • Cas Number : 1402240-25-8
  • Molecular Weight : 285.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart