CO1H324A6B5D CombiPhos Catalysts Inc

2-(Piperidon-1-yl)pyridine-5-boronic acid pinacol ester

  • Cas Number : 2223043-44-3
  • Molecular Weight : 302.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart