CO1H324A6B6F CombiPhos Catalysts Inc

2-(Cyclopentoxy)pyridine-5-boronic acid pinacol ester

  • Cas Number : 1432572-05-8
  • Molecular Weight : 289.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart