CO1H324A6B77 CombiPhos Catalysts Inc

2,3-Dibromopyridine-4-boronic acid pinacol ester

  • Cas Number : 1402237-95-9
  • Molecular Weight : 362.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart