CO1H324A6B7A CombiPhos Catalysts Inc

2-Methyl-3-bromopyridine-4-boronic acid pinacol ester

  • Cas Number : 1402172-60-4
  • Molecular Weight : 297.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart