CO1H324A6B86 CombiPhos Catalysts Inc

5-(n-Propyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 1884166-49-7
  • Molecular Weight : 247.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-propyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Inchi Code 1S/C14H22BNO2/c1-6-7-11-8-12(10-16-9-11)15-17-13(2,3)14(4,5)18-15/h8-10H,6-7H2,1-5H3
Inchi Key ZSUNHESFMXQOJS-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart