CO1H324A6B94 CombiPhos Catalysts Inc

5-(2-Methyl-4-fluorophenyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 1402227-29-5
  • Molecular Weight : 313.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart