CO1H324A6B97 CombiPhos Catalysts Inc

5-(4-Fluorophenyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 1105663-88-4
  • Molecular Weight : 299.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart