CO1H324A6B9C CombiPhos Catalysts Inc

5-(3, 5-Difluorophenyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 1402227-57-9
  • Molecular Weight : 317.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart