CO1H324A6BA7 CombiPhos Catalysts Inc

5-(2-Furyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223054-02-0
  • Molecular Weight : 271.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart