CO1H324A6BA8 CombiPhos Catalysts Inc

5-(3-Thienyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 1402240-75-8
  • Molecular Weight : 287.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart