CO1H324A6BAA CombiPhos Catalysts Inc

5-(2-Pyridyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223012-95-9
  • Molecular Weight : 282.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart