CO1H324A6BAF CombiPhos Catalysts Inc

5-(Pyrrolidino)pyridine-3-boronic acid pinacol ester

  • Cas Number : 1201644-49-6
  • Molecular Weight : 274.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart