CO1H324A6BB5 CombiPhos Catalysts Inc

5-(2-Methylimidazol-1-yl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 1402240-20-3
  • Molecular Weight : 285.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart