CO1H324A6BB9 CombiPhos Catalysts Inc

5-(Diethylamino)pyridine-3-boronic acid pinacol ester

  • Cas Number : 1257554-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart