CO1H324A6BCE CombiPhos Catalysts Inc

5-(Morpholino)pyridine-3-boronic acid pinacol ester

  • Cas Number : 1201644-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart