CO1H324A6BE3 CombiPhos Catalysts Inc

6-(n-Propoxy)pyrazine-2-boronic acid pinacol ester

  • Cas Number : 2223039-01-6
  • Molecular Weight : 264.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart