CO1H324A6BED CombiPhos Catalysts Inc

6-(Cyclopentoxy)pyrazine-2-boronic acid pinacol ester

  • Cas Number : 1416253-01-4
  • Molecular Weight : 290.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart