CO1H324A6BF0 CombiPhos Catalysts Inc

6-(N-Methylpiperazin-1-yl)pyrazine-2-boronic acid pinacol ester

  • Cas Number : 1186042-00-1
  • Molecular Weight : 304.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart