CO1H324A6BF2 CombiPhos Catalysts Inc

6-Ethylpyrazine-2-boronic acid pinacol ester

  • Cas Number : 1162262-41-0
  • Molecular Weight : 234.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart