CO1H324A6C0B CombiPhos Catalysts Inc

2-(sec-Butyl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223009-20-7
  • Molecular Weight : 261.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart