CO1H324A6C0E CombiPhos Catalysts Inc

3-Amino-pyridine-4-boronic acid pinacol ester

  • Cas Number : 1052714-68-7
  • Molecular Weight : 220.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart