CO1H324A6C1F CombiPhos Catalysts Inc

4-(tert-Butyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223012-99-3
  • Molecular Weight : 261.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart