CO1H324A6C21 CombiPhos Catalysts Inc

6-(4-Fluorophenyl)pyrazine-2-boronic acid pinacol ester

  • Cas Number : 1402172-96-6
  • Molecular Weight : 300.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart