CO1H324A6C2A CombiPhos Catalysts Inc

6-(3-Methoxyphenyl)pyrazine-2-boronic acid pinacol ester

  • Cas Number : 1402227-21-7
  • Molecular Weight : 312.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart