CO1H324A6C2E CombiPhos Catalysts Inc

5-Methyl-2-(ethyl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223045-66-5
  • Molecular Weight : 247.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart