CO1H324A6C35 CombiPhos Catalysts Inc

4-Bromo-3-methylpyridine-2-boronic acid pinacol ester

  • Cas Number : 2223044-13-9
  • Molecular Weight : 297.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart