CO1H324A6C3B CombiPhos Catalysts Inc

3-Cyano-6-methylpyridine-4-boronic acid pinacol ester

  • Cas Number : 2223027-55-0
  • Molecular Weight : 244.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart