CO1H324A6C3F CombiPhos Catalysts Inc

2,3-Dimethylpyridine-4-boronic acid pinacol ester

  • Cas Number : 1815616-21-7
  • Molecular Weight : 233.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart