CO1H324A6C41 CombiPhos Catalysts Inc

6-(4-Bromothiazol-2-yl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223030-30-4
  • Molecular Weight : 367.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart