CO1H324A6C43 CombiPhos Catalysts Inc

5-(5-Aminopyrazin-2-yl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223027-19-6
  • Molecular Weight : 298.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart