CO1H324A6C46 CombiPhos Catalysts Inc

5-(Thiazol-5-yl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223035-87-6
  • Molecular Weight : 288.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart