CO1H324A6C47 CombiPhos Catalysts Inc

6-(4-Fluoro-2-methylphenyl)pyridine-2-boronic acid pinacol ester

  • Cas Number : 1402227-30-8
  • Molecular Weight : 313.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart