CO1H324A6C4F CombiPhos Catalysts Inc

2-(n-Propyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223044-36-6
  • Molecular Weight : 247.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart