CO1H324A6C5F CombiPhos Catalysts Inc

2-(5-Pyrimidyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223039-26-5
  • Molecular Weight : 283.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart