CO1H324A6C61 CombiPhos Catalysts Inc

2-(3-Tolyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223035-97-8
  • Molecular Weight : 295.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart