CO1H324A6C66 CombiPhos Catalysts Inc

2-(2-Fluorophenyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223038-68-2
  • Molecular Weight : 299.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart