CO1H324A6C6B CombiPhos Catalysts Inc

2-(4-Trifluoromethylphenyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223029-08-9
  • Molecular Weight : 349.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart