CO1H324A6C73 CombiPhos Catalysts Inc

2-(2-Aminophenyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223028-73-5
  • Molecular Weight : 296.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart