CO1H324A6C92 CombiPhos Catalysts Inc

4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • Cas Number : 1220219-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart