CO1H324A6C93 CombiPhos Catalysts Inc

2-(4-(tert-butyl)phenyl)-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • Cas Number : 1080642-58-5
  • Molecular Weight : 337.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart