CO1H324A6C9E CombiPhos Catalysts Inc

2-(N-Boc-3,6-Dihydro-2H-pyridin-4-yl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2225172-91-6
  • Molecular Weight : 386.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart