CO1H324A6CA1 CombiPhos Catalysts Inc

2-[(5', 5'-Dimethyl)tetrahydrofuran-3-yl]pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223005-49-8
  • Molecular Weight : 303.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart